Anti-MKI67 Mouse Monoclonal Antibody [clone: MIB-1]
Catalog # ABNOMAB3545
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Marker of proliferation Ki-67
- Antigen symbol:MKI67
- Clonality:Monoclonal
- Clone:MIB-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:4288
- Antigen synonyms:MIB-1|MIB-|KIA|PPP1R105
- Storage buffer:phosphate buffer, pH 7,3 (0,09% sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Native purified MKI67 from human nuclear Ki-67 antigen.
- Size:5 ml
- Pk:5 ml
Specifications
About this item
Mouse monoclonal antibody raised against MKI67.
Type: Primary
Antigen: MKI67
Clonality: Monoclonal
Clone: MIB-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human