Anti-REN Mouse Monoclonal Antibody [clone: 244/1 RH12]
Catalog # ABNOMAB3694
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Renin
- Antigen symbol:REN
- Clonality:Monoclonal
- Clone:244/1 RH12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:5972
- Antigen synonyms:renin precursor|HNFJ2|angiotensinogenase|renal|angiotensin-forming enzyme
- Storage buffer:BBS
- Storage temperature:Store at 4 °C.
- Immunogen:Recombinant protein corresponding to human REN.
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant REN.
Type: Primary
Antigen: REN
Clonality: Monoclonal
Clone: 244/1 RH12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human