Anti-MKI67 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12127
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Marker of proliferation Ki-67
- Antigen symbol:MKI67
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:17345
- Antigen synonyms:MIB-1|MIB-|KIA|PPP1R105
- Amino acid number:1600 to 1700
- Storage buffer:Tris-citrate/phosphate buffer, pH 7.0-8.0 (0,1% sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to amino acids 1600-1700 of mouse Mki67.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Mki67.
Recommended Dilutions: Flow Cytometry: 1:100; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100-1:500
Type: Primary
Antigen: MKI67
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse