Order Entry
Italy
ContactUsLinkComponent
Anti-SPN Mouse Monoclonal Antibody (Biotin) [clone: MEM-59]
Anti-SPN Mouse Monoclonal Antibody (Biotin) [clone: MEM-59]
Catalog # ABNOMAB4548
Supplier:  Abnova
Anti-SPN Mouse Monoclonal Antibody (Biotin) [clone: MEM-59]
Catalog # ABNOMAB4548
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sialophorin
  • Antigen symbol:
    SPN
  • Clonality:
    Monoclonal
  • Clone:
    MEM-59
  • Conjugation:
    Biotin
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    6693
  • Antigen synonyms:
    LSN|CD43|GPL115|GALGP
  • Storage buffer:
    PBS, pH 7,2 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified SPN from human T lymphocytes.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against native SPN.

Recommended Dilutions: Flow Cytometry: 1:200

Type: Primary
Antigen: SPN
Clonality: Monoclonal
Clone: MEM-59
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human