Anti-CRYAA Mouse Monoclonal Antibody [clone: 1H3.B8]
ABNOMAB2178
: Abnova
- Antibody type:Primary
- Antigen name:Crystallin, alpha A
- Antigen symbol:CRYAA
- Clonality:Monoclonal
- Clone:1H3.B8
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1409
- Antigen synonyms:HSPB4|CTRCT9|CRYA1
- Storage buffer:PBS, pH 7,4
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CRYAA.
- Size:100 μg
- Pk:100 µl
Mouse monoclonal antibody raised against native CRYAA.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CRYAA
Clonality: Monoclonal
Clone: 1H3.B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat