Anti-CTRC Mouse Monoclonal Antibody
ABNOMAB2211
: Abnova
- Antibody type:Primary
- Antigen name:Chymotrypsin C
- Antigen symbol:CTRC
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG3
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:11330
- Antigen synonyms:ELA4|CLCR
- Storage buffer:PBS, pH 7,4
- Storage temperature:Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CTRC from human pancreas.
- Pk:100 µG
Mouse monoclonal antibody raised against native CTRC.
Type: Primary
Antigen: CTRC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human