Anti-ITGB2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-48]
Catalog # ABNOMAB4599
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Integrin Beta 2
- Antigen symbol:ITGB2
- Clonality:Monoclonal
- Clone:MEM-48
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:3689
- Antigen synonyms:LFA-1|CD18|MAC-1|integrin subunit beta 2|LAD|LCAMB|MFI7|MF17
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified ITGB2 from leukocytes of patient suffering from LGL-type leukaemia.
- Size:100 Reactions
- Pk:100 Tests
Specifications
About this item
Mouse monoclonal antibody raised against native ITGB2.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone: MEM-48
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human