Anti-CD14 Mouse Monoclonal Antibody (Biotin) [clone: MEM-18]
Catalog # ABNOMAB4613
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD14 Molecule
- Antigen symbol:CD14
- Clonality:Monoclonal
- Clone:MEM-18
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Primates,Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:929
- Antigen synonyms:monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- Storage buffer:PBS, pH 7,2 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified human CD14.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against native CD14.
Recommended Dilutions: Flow Cytometry: 1:1000
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: MEM-18
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-human Primates