Order Entry
Italy
ContactUsLinkComponent
Anti-CD3D Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-57]
Anti-CD3D Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-57]
Catalog # ABNOMAB4616
Supplier:  Abnova
Anti-CD3D Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-57]
Catalog # ABNOMAB4616
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD3 Molecule Delta
  • Antigen symbol:
    CD3D
  • Clonality:
    Monoclonal
  • Clone:
    MEM-57
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    915|916|917
  • Antigen synonyms:
    CD3-DELTA|IMD19|T3D
  • Storage buffer:
    PBS (0,2% BSA, 15 mM sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified CD3 from human thymocytes and T lymphocytes.
  • Size:
    100 Reactions
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against native CD3.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD3D
Clonality: Monoclonal
Clone: MEM-57
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human