Anti-CD53 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-53]
ABNOMAB4639
: Abnova
- Antibody type:Primary
- Antigen name:Leukocyte surface antigen CD53
- Antigen symbol:CD53
- Clonality:Monoclonal
- Clone:MEM-53
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:963
- Antigen synonyms:MOX44|TSPAN25|CD53 molecule
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD53 from leukocytes of patient suffering from LGL-type leukaemia.
- Size:100 Reactions
- Pk:100 µg
Mouse monoclonal antibody raised against native CD53.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: CD53
Clonality: Monoclonal
Clone: MEM-53
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human