Anti-SLC9A3 Rabbit Polyclonal Antibody
ABNOPAB23047
: Abnova
- Antibody type:Primary
- Antigen name:Solute Carrier Family 9 Member 3
- Antigen symbol:Solute carrier family 9, member 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:6550
- Antigen synonyms:isoform 3|Solute carrier family 9 sodium/hydrogen exchanger|NHE3|Solute carrier family 9 member 3|SLC9A 3|Slc9a3|MGC126718|Sodium/hydrogen exchanger|member 3|NHE-3|MGC126720.|Sodium/hydrogen exchanger 3|SL9A3_HUMAN|NHE 3|Na+/H+ exchanger 3|Sodium|apical epithelial
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:DNPVFSPDEALDRSLLARLPPWLSPGETVVPSQRARTQIPYSPGTFCRLMPFRLSSKSVDSFLQADGPEERPPAALPEST
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human SLC9A3.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against recombinant SLC9A3.
Recommended Dilutions: Immunohistochemistry: 1:200-1:500
Type: Primary
Antigen: SLC9A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human