Order Entry
Italy
ContactUsLinkComponent
Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9]
Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9]
Catalog # ABNOMAB5620
Supplier:  Abnova
Anti-ITGAX Mouse Monoclonal Antibody [clone: 3.9]
Catalog # ABNOMAB5620
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Integrin Alpha 10
  • Antigen symbol:
    ITGA10
  • Clonality:
    Monoclonal
  • Clone:
    3.9
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3687
  • Antigen synonyms:
    integrin alpha-X|integrin aX|ITGAX|AI449405|CD11 antigen-like family member C|N418|Cr4|Cd11c
  • Storage buffer:
    100 mM BBS, pH 8,2
  • Storage temperature:
    Store at 4 °C.
  • Immunogen:
    Native purified ITGAX from human rheumatoid synovial fluid cells and fibronectin purified human monocytes.
  • Purification:
    Ammonium sulfate precipitation
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native ITGAX.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: 3.9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human