Anti-FCGR2B Mouse Monoclonal Antibody [clone: 7.3]
Catalog # ABNOMAB6930
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fc fragment of IgG, low affinity 2B, receptor (CD32)
- Antigen symbol:FCGR2B
- Clonality:Monoclonal
- Clone:7.3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:2213
- Antigen synonyms:CD32|IGFR2|FCG2|FCGR2|CD32B
- Storage buffer:50 mM sodium phosphate buffer, 100 mM potassium Chloride, 150 mM NaCl, pH 7,5 (0,5 mg/mL gentamicsulfate)
- Storage temperature:Store at 4 °C.
- Immunogen:Native purified FCGR2B from human K-562 tumor cells and L cells transfected with human Fc gamma RII.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native FCGR2B.
Type: Primary
Antigen: FCGR2B
Clonality: Monoclonal
Clone: 7.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human