Anti-V5 Tag Monoclonal Antibody [clone: E10]
Catalog # ABNOMAB8756
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:V5 tag
- Antigen symbol:V5
- Clonality:Monoclonal
- Clone:E10
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,4 (0,05% sodium azide)
- Sequence:GKPIPNPLLGLDST
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to V5 tag.
- Purification:Protein A purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant V5 tag.
Recommended Dilutions: ELISA: 1:5000-10000; Western Blot: 1:1000; Immunohistochemistry: 1:50-1:100
Type: Primary
Antigen: V5 Tag
Clonality: Monoclonal
Clone: E10
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: