Anti-CK4, C5, C6, C8, C10, C13, and C18. Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C-11]
ABNOMAB5131
: Abnova
- Antibody type:Primary
- Antigen name:Cytokeratin 4, 5, 6, 8, 10, 13 And 18
- Antigen symbol:CK4, C5, C6, C8, C10, C13, and C18.
- Clonality:Monoclonal
- Clone:C-11
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Mammals,Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified cytokeratin from Keratin-enriched preparation from human epidermoid carcinoma cell line A431.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native cytokeratin.
Recommended Dilutions: Flow Cytometry: 5 ?g/ml
Type: Primary
Antigen: CK4, C5, C6, C8, C10, C13, and C18.
Clonality: Monoclonal
Clone: C-11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mammals