Anti-SETD1A Rabbit Polyclonal Antibody
Catalog # ABNOPAB14083
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:SET Domain Containing 1A
- Antigen symbol:SETD1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Serum
- Gene ID:233904
- Antigen synonyms:KMT2F|BC010250|Nsccn1|mNSC1|mKIAA0339
- Storage buffer:0,05% sodium azide
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A mixture of synthetic peptides (conjugated with KLH), of which two correspond to N-terminus and one to internal region of mouse Setd1a.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Setd1a.
Recommended Dilutions: ELISA: 1:100; Western Blot: 1:500
Type: Primary
Antigen: SETD1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse