Anti-Hepatitis B Virus Surface Antigen surface Mouse Monoclonal Antibody [clone: S 38]
Catalog # ABNOMAB5400
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Hepatitis B Virus Surface Antigen surface
- Clonality:Monoclonal
- Clone:S 38
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Virus
- Cross adsorption:No
- Format:Liquid
- Form:Serum
- Storage buffer:Serum
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native HBsAg from human sera pool.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native hepatitis B virus surface antigen.
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: S 38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus