Anti-Influenza A (H3N2) Goat Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # ABNOPAB14280
Supplier: Abnova
Specifications
- Antigen name:Influenza A (H3N2)
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Goat
- ImmunoFluorescence:Yes
- Reactivity:Virus
- Cross adsorption:No
- Format:Liquid
- Storage buffer:10 mM PBS, pH 7,2 (10 mg/mL BSA, 0,1% sodium azide)
- Storage temperature:Store in the dark at 4 °C. For long term storage store at –20 °C. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Influenza A Strain Texas 1/77 (H3N2).
- Pk:1 mL
Specifications
About this item
Goat polyclonal antibody raised against Influenza A, strain Texas 1/77 (H3N2).
Type:
Antigen: Influenza A (H3N2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Virus