Anti-ZNF76 Rabbit Polyclonal Antibody
Catalog # ABNOPAB17363
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 76
- Antigen symbol:ZNF76
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:7629
- Antigen synonyms:Zfp523|D6S229E|ZNF523
- Storage buffer:PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human ZNF76.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of ZNF76.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; ELISA: 1:10000
Type: Primary
Antigen: ZNF76
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human