Anti-NKX2-1 Rabbit Monoclonal Antibody [clone: G21-G]
Catalog # ABNOMAB9783
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NK2 homeobox 1
- Antigen symbol:NKX2-1
- Clonality:Monoclonal
- Clone:G21-G
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:7080
- Antigen synonyms:TTF-1|BCH|NKX2A|TEBP|NKX2.1|TTF1|BHC|NK-2|T/EBP|TITF1
- Storage buffer:20 mM Tris-HCl, pH 8.0 (20 mg/mL BSA, 0,05% sodium azide)
- Storage temperature:Store at 4 °C.
- Immunogen:A synthetic peptide corresponding to N-terminus of human NKX2-1.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit monoclonal antibody raised against synthetic peptide of NKX2-1.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:100-1:200
Type: Primary
Antigen: NKX2-1
Clonality: Monoclonal
Clone: G21-G
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse