Anti-TP73 Rabbit Polyclonal Antibody
ABNOPAB5789
: Abnova
- Antibody type:Primary
- Antigen name:Tumor protein 73
- Antigen symbol:TP73
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:7161
- Antigen synonyms:p53-like transcription factor|P73|TRP73|p53-related protein
- Modification:phospho-Tyr99
- Storage buffer:PBS, 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding Y99 of human TP73.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic phosphopeptide of TP73.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: TP73
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat