Anti-SLC39A7 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19352
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 39 Member A7
- Antigen symbol:Solute carrier family 39, member 7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:7922
- Antigen synonyms:HKE4|H2-KE4|D6S115E|Zinc transporter SLC39A7|D6S2244E|RING5|Histidine-rich membrane protein Ke4|KE4|ZIP7
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 17 amino acids near N-terminus of human SLC39A7.
- Purification:Peptide affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody rasied against synthetic peptide of SLC39A7.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunohistochemistry: 2.5 ?g/ml
Type: Primary
Antigen: SLC39A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat