Anti-IL1A Mouse Monoclonal Antibody [clone: 5G3]
ABNOMAB9932
: Abnova
- Antibody type:Primary
- Antigen name:Interleukin 1 alpha
- Antigen symbol:IL1A
- Clonality:Monoclonal
- Clone:5G3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3552
- Antigen synonyms:Hematopoietin-1|IL-1 alpha|pro-interleukin-1-alpha|IL1A|preinterleukin 1 alpha|hematopoietin-1|interleukin-1 alpha
- Storage buffer:Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human IL1A.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against full length recombinant IL1A.
Recommended Dilutions: Western Blot: 1:1000; Immunohistochemistry: 1:10
Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone: 5G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human