Anti-ATP13A1 Goat Polyclonal Antibody
Catalog # ABNOPAB7520
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ATPase type 13A1
- Antigen symbol:ATP13A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:57130
- Antigen synonyms:CGI-152|ATP13A
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-REAQARSPEKQEQ
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human ATP13A1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of ATP13A1.
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: ATP13A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human