Order Entry
Italy
ContactUsLinkComponent
Anti-17-alpha-Trenbolone Polyclonal Antibody
Anti-17-alpha-Trenbolone Polyclonal Antibody
  ABNOPAB8204
 :  Abnova
Anti-17-alpha-Trenbolone Polyclonal Antibody
  ABNOPAB8204
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    17-alpha-Trenbolone
  • Antigen symbol:
    17-alpha-Trenbolone
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    20 mM phosphate buffer, 150 mM NaCl, pH 7,2 (0,09% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    17 alpha-Trenbolone conjugated with BTG.
  • Pk:
    1 mg

 

 

Sheep polyclonal antibody raised against 17 alpha-Trenbolone.

Recommended Dilutions: Enzyme Immunoassay (0.5 ?g/ml

Type: Primary
Antigen: 17-alpha-Trenbolone
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: