Order Entry
Italy
ContactUsLinkComponent
Anti-17-beta-Testosterone-long linker Polyclonal Antibody
Anti-17-beta-Testosterone-long linker Polyclonal Antibody
Catalog # ABNOPAB8213
Supplier:  Abnova
Anti-17-beta-Testosterone-long linker Polyclonal Antibody
Catalog # ABNOPAB8213
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    17-beta-Testosterone-long linker
  • Antigen symbol:
    17-beta-Testosterone-long linker
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    20 mM phosphate buffer, 150 mM NaCl, pH 7,2 (0,09% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    17 beta-Testosterone-long linker-BSA.
  • Pk:
    1 mg

Specifications

About this item

Sheep polyclonal antibody raised against 17 beta-Testosterone-long linker.

Recommended Dilutions: Enzyme Immunoassay (2.5 ?g/ml

Type: Primary
Antigen: 17-beta-Testosterone-long linker
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: