Anti-SLC1A5 Rabbit Polyclonal Antibody
Catalog # ABNOPAB26061
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 1 (neutral amino acid transporter), member 5
- Antigen symbol:Solute carrier family 1, member 5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:6510
- Antigen synonyms:RDRC|M7VS1|AAAT|ATBO|R16|ASCT2|M7V1
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 15 amino acids at C-terminus of human SLC1A5.
- Purification:Immunoaffinity chromatography
- Size:50 μg
- Pk:50 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SLC1A5.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2.5 ?g/ml
Type: Primary
Antigen: SLC1A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human