Anti-GPR146 Rabbit Polyclonal Antibody
Catalog # ABNOPAB26352
Nome del fornitore: Abnova
Specifiche
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 146
- Antigen symbol:GPR146
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Hamster,Gorilla,Human,Rat,Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:115330
- Antigen synonyms:PGR8|GP146_HUMAN|G protein coupled receptor 146|GPR146|Probable G-protein coupled receptor 146|GPCR GPR146.|G-protein coupled receptor PGR8
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 19 amino acids at 2nd cytoplasmic domain of human GPR146.
- Purification:Immunoaffinity chromatography
- Size:50 μg
- Pk:50 µg
Specifiche
Informazioni su questo articolo
Rabbit polyclonal antibody raised against synthetic peptide of GPR146.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 25 ?g/ml
Type: Primary
Antigen: GPR146
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Hamster, Human, Mouse, Rat