Order Entry
Italy
ContactUsLinkComponent
 
Mouse IgG1 Isotype Control [clone: MOPC-21] (violetFluor® 450)
  75-4714-U100
Mouse IgG1 Isotype Control [clone: MOPC-21] (violetFluor® 450)
  75-4714-U100

 

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-21
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Storage temperature:
    +2 °C to +8 °C, protected from light
  • Pk:
    100 µG

 

 

The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Type: Control
Antigen: MOPC-21
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: violetFluor™ 450
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: