Order Entry
Italy
ContactUsLinkComponent
Anti-TOX2 Rabbit Polyclonal Antibody
Anti-TOX2 Rabbit Polyclonal Antibody
Catalog # ABGEAP19478C
Supplier:  Abgent
Anti-TOX2 Rabbit Polyclonal Antibody
Catalog # ABGEAP19478C
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    TOX high mobility group box family member 2
  • Antigen symbol:
    TOX2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    84969
  • Antigen synonyms:
    C20orf100|dJ1108D11.2|dJ495O3.1|GCX-1|GCX1
  • Amino acid number:
    286 to 312
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    51.604 kDa
  • Sequence:
    KQSSPDQGETKSC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.42
  • Shipping temperature:
    4 °C
  • Immunogen:
    This TOX2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 286-312 amino acids from the Central region of human TOX2.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: TOX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human