Anti-P3H4 Rabbit Polyclonal Antibody
ABGEAP13238B
: Abgent
- Antibody type:Primary
- Antigen name:prolyl 3-hydroxylase family member 4 (non-enzymatic)
- Antigen symbol:P3H4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Gene ID:10609
- Antigen synonyms:SC65|LEPREL4|NO55|NOL55
- Amino acid number:361 to 390
- Storage buffer:pH 7,4
- Molecular weight:50.381 kDa
- Sequence:YLQSDDEMELEETEPC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.37
- Shipping temperature:4 °C
- Immunogen:This SC65 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 361-390 amino acids from the C-terminal region of human SC65.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: P3H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human