Anti-PDCD6IP Rabbit Polyclonal Antibody
Catalog # ABGEAP19973C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Programmed Cell Death 6 Interacting Protein
- Antigen symbol:PDCD6IP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:10015
- Antigen synonyms:ALIX|AIP1|HP95|DRIP4
- Amino acid number:541 to 570
- Storage buffer:pH 7,4
- Molecular weight:96.023 kDa
- Sequence:LSNLDEVKKEREGLEC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:1.010
- Shipping temperature:4 °C
- Immunogen:This PDCD6IP antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 541-570 amino acids from the Central region of human PDCD6IP.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PDCD6IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human