Anti-ZSCAN4 Rabbit Polyclonal Antibody
Catalog # ABGEAP18335A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Zinc finger and SCAN domain containing 4
- Antigen symbol:ZSCAN4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Gene ID:201516
- Antigen synonyms:ZNF494
- Amino acid number:94 to 120
- Storage buffer:pH 7,4
- Molecular weight:48.957 kDa
- Sequence:KSSGKNLERFIEC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.41
- Shipping temperature:4 °C
- Immunogen:This ZSCAN4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 94-120 amino acids from the N-terminal region of human ZSCAN4.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ZSCAN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human