Order Entry
Italy
ContactUsLinkComponent
Anti-TFAP2E Rabbit Polyclonal Antibody
Anti-TFAP2E Rabbit Polyclonal Antibody
Catalog # ABGEAZB10019A
Supplier:  Abgent
Anti-TFAP2E Rabbit Polyclonal Antibody
Catalog # ABGEAZB10019A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Transcription Factor AP-2 epsilon
  • Antigen symbol:
    TFAP2E
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Zebrafish
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    393794
  • Antigen synonyms:
    zgc:65882|zgc:77338
  • Amino acid number:
    304 to 330
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    45.514 kDa
  • Sequence:
    RQTEPDQLPTRRC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.33
  • Shipping temperature:
    4 °C
  • Immunogen:
    This DANRE tfap2e antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 304-330 amino acids from the C-terminal region of DANRE tfap2e.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: TFAP2E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Zebrafish