Anti-POLE2 Rabbit Polyclonal Antibody
ABGEAP20396B
: Abgent
- Antibody type:Primary
- Antigen name:Polymerase (DNA directed), epsilon 2, accessory subunit
- Antigen symbol:POLE2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Gene ID:5427
- Antigen synonyms:DPE2
- Amino acid number:495 to 523
- Storage buffer:pH 7,4
- Molecular weight:59.537 kDa
- Sequence:KVFYPSNKTVEDSKC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:2.400
- Shipping temperature:4 °C
- Immunogen:This POLE2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 495-523 amino acids from the C-terminal region of human POLE2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: POLE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human