Anti-WAS Rabbit Polyclonal Antibody
Catalog # ABGEAP19544C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Wiskott-Aldrich syndrome
- Antigen symbol:WAS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:7454
- Antigen synonyms:THC1|IMD2|SCNX|WASP|THC|WASPA
- Amino acid number:205 to 234
- Storage buffer:pH 7,4
- Molecular weight:52.913 kDa
- Sequence:PSPADKKRSGKKKISC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.64
- Shipping temperature:4 °C
- Immunogen:This WAS antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 205-234 amino acids from the Central region of human WAS.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: WAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human