Order Entry
Italy
ContactUsLinkComponent
Anti-UGT2B10 Rabbit Polyclonal Antibody
Anti-UGT2B10 Rabbit Polyclonal Antibody
  ABGEAP17222B
 :  Abgent
Anti-UGT2B10 Rabbit Polyclonal Antibody
  ABGEAP17222B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    UDP glucuronosyltransferase 2 family, polypeptide B10
  • Antigen symbol:
    UGT2B10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    7365
  • Antigen synonyms:
    UDPGT2B10
  • Amino acid number:
    431 to 458
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    60.774 kDa
  • Sequence:
    RIQHDQPVKPLDRC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.45
  • Shipping temperature:
    4 °C
  • Immunogen:
    This UGT2B10 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 431-458 amino acids from the C-terminal region of human UGT2B10.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: UGT2B10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human