Anti-ZNF37A Rabbit Polyclonal Antibody
Catalog # ABGEAP18996A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:zinc finger protein 37A
- Antigen symbol:ZNF37A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Gene ID:7587
- Antigen synonyms:ZNF37|KOX21
- Amino acid number:71 to 100
- Storage buffer:pH 7,4
- Molecular weight:65.418 kDa
- Sequence:RNYSIMKFNEFNKGGC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5
- Shipping temperature:4 °C
- Immunogen:This ZNF37A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 71-100 amino acids from the N-terminal region of human ZNF37A.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ZNF37A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human