Order Entry
Italy
ContactUsLinkComponent
Anti-F1LVT0 Rabbit Polyclonal Antibody
Anti-F1LVT0 Rabbit Polyclonal Antibody
  ABGEAP3881A
 :  Abgent
Anti-F1LVT0 Rabbit Polyclonal Antibody
  ABGEAP3881A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen symbol:
    F1LVT0
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Modification:
    phospho-Ser284
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    159.796 kDa
  • Sequence:
    SLLQEQSBLQEKYNC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.3
  • Shipping temperature:
    4 °C
  • Immunogen:
    This rat CCNB3 Antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S284 of rat CCNB3.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: F1LVT0
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat