Anti-SSX11 Rabbit Polyclonal Antibody
Catalog # ABGEAP19075A
Supplier: Abgent
Jump to:
Product Details & Documents
Type: Primary
Antigen: SSX11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Documents
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Protein A & Peptide affinity purification
- Antibody type:Primary
- Epitope:N-terminal
- Amino acid number:1 to 30
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Host:Rabbit
- Sequence:KRARDDAQIPEKC
- Concentration:0.52
- Shipping temperature:4 °C
- Cat. No.:ABGEAP19075A
- Reactivity:Human
- Immunogen:This SSX11 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human SSX11.
- Conjugation:Unconjugated
- Antigen symbol:SSX11
- Storage buffer:pH 7,4
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



