Anti-CYP1A Rabbit Polyclonal Antibody [clone: CP-226]
Supplier: Cayman Chemical
Certificates
About this item
Type: Primary
Antigen: CYP1A
Clonality: Polyclonal
Clone: CP-226
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Atlantic Cod, Carp, Lemon sole, mudfish, Rainbow Trout, Salmon, Turbot
Specifications
- Catalog No:
- CAYM173135-100
- CAYM173135-500
- Antigen symbol:
- CYP1A
- CYP1A
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- CP-226
- CP-226
- Reactivity:
- Lemon sole, Carp, Mudfish, Atlantic cod, Rainbow trout, Turbot, Salmon
- Lemon sole, Carp, Mudfish, Atlantic cod, Rainbow trout, Turbot, Salmon
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- rainbow trout CYP1A amino acid sequence 190-204 and 282-296, coupled to the carrier protein KLH
- rainbow trout CYP1A amino acid sequence 190-204 and 282-296, coupled to the carrier protein KLH
- Amino acid number:
- 190 to 204 282 to 296
- 190 to 204 282 to 296
- Purification:
- purified
- purified
- Cross adsorption:
- No
- No
- Storage buffer:
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- 0,1 M sodium phosphate, pH 7,4, containing 0,15 M sodium chloride, 12.3 mM sodium azide, and 1% BSA
- Storage temperature:
- +4 °C
- +4 °C
- Shipping temperature:
- 4 °C
- 4 °C
Specifications
Frequently Bought Together