Order Entry
Italy
ContactUsLinkComponent
Anti-COX-2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CX229]
Anti-COX-2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CX229]
  CAYM160113-1
Anti-COX-2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CX229]
  CAYM160113-1

 

  • Antibody type:
    Primary
  • Antigen symbol:
    COX-2
  • Clonality:
    Monoclonal
  • Clone:
    CX229
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Sheep,
    Human
  • Cross adsorption:
    No
  • Amino acid number:
    580 to 599
  • Sequence:
    NASSSRSGLDDINPTVLLKE
  • Storage temperature:
    +4 °C
  • Shipping temperature:
    4 °C
  • Immunogen:
    human COX-2 amino acids 580-599
  • Purification:
    purified
  • Size:
    1 Ea
  • Pk:
    1

 

 

Type: Primary
Antigen: COX-2
Clonality: Monoclonal
Clone: CX229
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Sheep