Order Entry
Italy
ContactUsLinkComponent
Anti-HCN1 Mouse Monoclonal Antibody [clone: S70-28]
Anti-HCN1 Mouse Monoclonal Antibody [clone: S70-28]
  CAYM13705-100
Anti-HCN1 Mouse Monoclonal Antibody [clone: S70-28]
  CAYM13705-100

 

  • Antibody type:
    Primary
  • Antigen symbol:
    HCN1
  • Clonality:
    Monoclonal
  • Clone:
    S70-28
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    No
  • Amino acid number:
    778 to 910
  • Storage buffer:
    PBS, pH 7,4, containing 50% glycerol and 0,09% sodium azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    fusion protein amino acids 778-910 (cytoplasmic C-terminus) of rat HCN1
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: HCN1
Clonality: Monoclonal
Clone: S70-28
Conjugation: unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat