Anti-ANGPTL3 Mouse Monoclonal Antibody [clone: Kairos-37]
Nome del fornitore: Cayman Chemical
Certificati
Informazioni su questo articolo
Type: Primary
Antigen: ANGPTL3
Clonality: Monoclonal
Clone: Kairos-37
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifiche
- Catalog No:
- CAYM10805-100
- CAYM10805-50
- Antigen symbol:
- Aminomethyltransferase
- ANGPTL3
- Antigen name:
- Angiopoietin-like 3
- Angiopoietin-like 3
- Antigen synonyms:
- ANGPTL3
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- Kairos-37
- Kairos-37
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- recombinant human ANGPTL3
- recombinant human ANGPTL3
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,4
- PBS, pH 7,4
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- –20 °C
- –20 °C
Specifiche
Frequently Bought Together