Order Entry
Italy
ContactUsLinkComponent
 
Anti-RSK2 Rabbit Polyclonal Antibody
Catalog # CAYM10009411-1
Supplier:  Cayman Chemical
Anti-RSK2 Rabbit Polyclonal Antibody
Catalog # CAYM10009411-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    RSK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    near C-terminus
  • Cross adsorption:
    No
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    peptide corresponding to amino acid residues from the C-terminal region of rat RSK2
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: RSK2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: near C-terminus
Host: Rabbit
Isotype:
Reactivity: Rat