Anti-DsRed Mouse Monoclonal Antibody (Unconjugated) [clone: 8E5.G7]
Catalog # ROCK200-301-379
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Tag
- Antigen symbol:DsRed
- Clonality:Monoclonal
- Clone:8E5.G7
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-RFP monoclonal antibody is a Red Fluorescent Protein (RFP) fusion protein corresponding to the full length amino acid sequence (234aa) derived from the mushroom polyp coral Discosoma.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-RFP antibodies is designed to detect Red Fluorescent Protein and its variants. This antibody can be used to detect RFP by ELISA (sandwich or capture) for the direct binding of antigen.
Recommended Dilutions: ELISA: 1:20000-1:50000; Immunofluorescence: User Optimized; Flow Cytometry: User Optimized; Western Blot: 1:1000-1:3000; Immunoprecipitation: User Optimized
Type: Tag
Antigen: DsRed
Clonality: Monoclonal
Clone: 8E5.G7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a kappa
Reactivity: