Anti-TPH1 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-E49
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:tryptophan hydroxylase 1
- Antigen symbol:TPH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:24848
- Antigen synonyms:Tph
- Modification:phospho-Ser260
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-Tryptophan Hydroxylase pS260 Antibody was produced in rabbit by repeated immunizations with synthetic phospho-peptide corresponding to amino acid residues surrounding Ser260 of Tryptophan Hydroxylase.
- Purification:affinity-purified
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-Tryptophan Hydroxylase pS260 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of 55 kDa in size corresponding to typtophan hydroxylase Protein phosphorylated at Ser 260.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: TPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat