Anti-SCN9A Mouse Monoclonal Antibody (Unconjugated) [clone: S68-6]
ROCK200-301-G21
- Antibody type:Primary
- Antigen name:sodium channel, voltage gated, type IX alpha subunit
- Antigen symbol:SCN9A
- Clonality:Monoclonal
- Clone:S68-6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:6335
- Antigen synonyms:ETHA|GEFSP7|HSAN2D|SFNP|PN1|FEB3B|NENA|Nav1.7|NE-NA
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Concentration:1 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:Nav1.7 Antibody was produced in mice by repeated immunizations raised against a fusion Protein corresponding to the cytoplamsic C-terminus region of human Nav1.7.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Anti-NAV1.7 Antibody is suitable for use in WB, IP, and Immunofluorescence microscopy. Expect a band approximately ~230kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: SCN9A
Clonality: Monoclonal
Clone: S68-6
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat, Human