Order Entry
Italy
ContactUsLinkComponent
 
Anti-SCN1A Mouse Monoclonal Antibody (Unconjugated) [clone: S74-71]
Catalog # ROCK200-301-G18
undefined
Anti-SCN1A Mouse Monoclonal Antibody (Unconjugated) [clone: S74-71]
Catalog # ROCK200-301-G18

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    sodium channel, voltage-gated, type I, alpha
  • Antigen symbol:
    SCN1A
  • Clonality:
    Monoclonal
  • Clone:
    S74-71
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    20265
  • Antigen synonyms:
    B230332M13|Nav1.1
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
  • Storage temperature:
    –20 °C
  • Concentration:
    1 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Nav1.1 Antibody was produced in mice by repeated immunizations raised against a fusion Protein corresponding to the cytoplamsic C-terminus region of rat Nav1.1.
  • Purification:
    Protein G purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Anti-Nav1.1 Antibody is suitable for use in WB, IP, and IHC. Expect a band approximately ~220kDa on specific lysates. Epitope mapped within amino acids 690 to 715. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: SCN1A
Clonality: Monoclonal
Clone: S74-71
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat