Anti-NELFB Mouse Monoclonal Antibody (Unconjugated) [clone: F7E4]
ROCK200-301-D18
- Antibody type:Primary
- Antigen name:Negative elongation factor complex member B
- Antigen symbol:NELFB
- Clonality:Monoclonal
- Clone:F7E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:25920
- Antigen synonyms:COBRA1|NELF-B
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-COBRA1 Monoclonal Antibody was produced in mouse by repeated immunizations with recombinant full length human COBRA1 purified from E.coli.
- Purification:Protein G purified
- Size:100 µl
- Pk:100 µl
Anti-COBRA 1 antibody is suitable for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 62 kDa in size corresponding to COBRA1 Proteins in neonatal Rat brain lysate.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000
Type: Primary
Antigen: NELFB
Clonality: Monoclonal
Clone: F7E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat, Human